Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casr
Entrez Gene ID
12374
Gene Name
calcium-sensing receptor
Gene Aliases
CaR, Gprc2a
Chromosome Location
Chr.16: 36490585 - 36562141 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013803.2 NP_038831.2 5 - 6 2129 56
GenBank mRNA AF068900.1 - 1 - 2 89 56
AF110178.1 - 5 - 6 2130 56
AF128842.1 - 4 - 5 1827 56
AF159565.1 - 2 - 3 815 56
AK029770.1 - 2 - 3 718 56

View Full Product Details