Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Opn4
Entrez Gene ID
30044
Gene Name
opsin 4 (melanopsin)
Gene Aliases
1110007J02Rik, Gm533
Chromosome Location
Chr.14: 34590618 - 34600142 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128599.1 NP_001122071.1 8 - 9 1434 58
NM_013887.2 NP_038915.1 8 - 9 1434 58
GenBank mRNA AF147789.1 - 8 - 9 1434 58
BC139827.1 - 8 - 9 1375 58
DQ278263.1 - 4 - 5 597 58
EU303117.1 - 8 - 9 1250 58
EU303118.1 - 8 - 9 1250 58

View Full Product Details