The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024853.1 | NP_001020024.1 | 4 - 5 | 375 | 95 |
NM_001024854.1 | NP_001020025.1 | 2 - 3 | 191 | 95 | |
NM_001286203.2 | NP_001273132.1 | 3 - 4 | 277 | 95 | |
NM_013896.3 | NP_038924.1 | 4 - 5 | 413 | 95 | |
GenBank mRNA | AF150101.1 | - | 3 - 4 | 236 | 95 |
AI119857.1 | - | 4 - 5 | 394 | 95 | |
AK018764.1 | - | 2 - 3 | 191 | 95 | |
AK165848.1 | - | 2 - 3 | 192 | 95 | |
AK172179.1 | - | 3 - 4 | 287 | 95 | |
BC024370.1 | - | 4 - 5 | 386 | 95 | |
CK626183.1 | - | 2 - 3 | 173 | 95 | |
CO799389.1 | - | 3 - 4 | 277 | 95 | |
CO809703.1 | - | - | 344 | 95 | |
DV652207.1 | - | 4 - 5 | 352 | 95 |