Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hnf4g
Entrez Gene ID
30942
Gene Name
hepatocyte nuclear factor 4, gamma
Gene Aliases
NR2A2
Chromosome Location
Chr.3: 3508030 - 3659800 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013920.2 NP_038948.1 7 - 8 1075 97
GenBank mRNA AJ242626.1 - 7 - 8 1027 97
AK143816.1 - 7 - 8 1076 97
BC125519.1 - 7 - 8 897 97

View Full Product Details