The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015732.4 | NP_056547.3 | 4 - 5 | 1408 | 70 |
GenBank mRNA | AF073788.1 | - | 4 - 5 | 1273 | 70 |
AF205889.1 | - | 3 - 4 | 1113 | 70 | |
AK049192.1 | - | 4 - 5 | 1352 | 70 | |
AK142333.1 | - | 4 - 5 | 1410 | 70 | |
BC057338.1 | - | 4 - 5 | 1391 | 70 |