Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcn2
Entrez Gene ID
21452
Gene Name
transcobalamin 2
Gene Aliases
AW208754, Tcn-2
Chromosome Location
Chr.11: 3917077 - 3932078 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130458.1 NP_001123930.1 3 - 4 454 75
NM_001130459.1 NP_001123931.1 3 - 4 423 75
NM_015749.3 NP_056564.1 3 - 4 457 75
GenBank mRNA AF090686.1 - 2 - 3 384 75
AK133707.1 - 3 - 4 419 75
AK143761.1 - 3 - 4 425 75
AK147176.1 - 3 - 4 428 75
AK161618.1 - 3 - 4 424 75
AK170401.1 - 3 - 4 443 75
BC003720.1 - 2 - 3 330 75

View Full Product Details