The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015814.2 | NP_056629.1 | 2 - 3 | 329 | 83 |
GenBank mRNA | AF177400.1 | - | 2 - 3 | 316 | 83 |
AJ243964.1 | - | 2 - 3 | 332 | 83 | |
AK004853.1 | - | 2 - 3 | 329 | 83 | |
AK013054.2 | - | 1 - 2 | 267 | 83 | |
AK013622.1 | - | 1 - 2 | 241 | 83 | |
BC046304.1 | - | 1 - 2 | 244 | 83 | |
BC050934.1 | - | 1 - 2 | 226 | 83 |