Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Reck
Entrez Gene ID
53614
Gene Name
reversion-inducing-cysteine-rich protein with kazal motifs
Gene Aliases
St15, mRECK
Chromosome Location
Chr.4: 43875530 - 43944806 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016678.2 NP_057887.2 2 - 3 236 72
GenBank mRNA AB006960.1 - 2 - 3 166 72
AK042266.1 - 2 - 3 237 72
AK143657.1 - 2 - 3 214 72
AK157885.1 - 2 - 3 224 72
BC053025.1 - 2 - 3 175 72
BC132197.1 - 2 - 3 264 72
BC138065.1 - 2 - 3 264 72
BC145133.1 - 2 - 3 264 72

View Full Product Details