The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016681.3 | NP_057890.1 | 12 - 13 | 1427 | 74 |
GenBank mRNA | AF086905.1 | - | 11 - 12 | 1388 | 74 |
AK044913.1 | - | 12 - 13 | 1428 | 74 | |
AK170235.1 | - | 11 - 12 | 1371 | 74 | |
BC056617.1 | - | 12 - 13 | 1451 | 74 |