Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map3k14
Entrez Gene ID
53859
Gene Name
mitogen-activated protein kinase kinase kinase 14
Gene Aliases
Nik, aly
Chromosome Location
Chr.11: 103219764 - 103267401 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016896.3 NP_058592.1 5 - 6 1239 82
GenBank mRNA AF143094.1 - 4 - 5 1156 82
AK036200.1 - 5 - 6 1240 82

View Full Product Details