The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163671.1 | NP_001157143.1 | 10 - 11 | 1327 | 68 |
NM_001163672.1 | NP_001157144.1 | 10 - 11 | 1327 | 68 | |
NM_016961.3 | NP_058657.1 | 10 - 11 | 1327 | 68 | |
NM_207692.2 | NP_997575.2 | 10 - 11 | 1327 | 68 | |
GenBank mRNA | AB005664.1 | - | 9 - 10 | 1101 | 68 |
AF052466.1 | - | 9 - 10 | 1063 | 68 | |
AF052467.1 | - | 9 - 10 | 1063 | 68 | |
AF052468.1 | - | 9 - 10 | 1063 | 68 | |
AF052469.1 | - | 9 - 10 | 1063 | 68 | |
AK031959.1 | - | 10 - 11 | 1326 | 68 | |
AK031974.1 | - | 8 - 9 | 881 | 68 | |
AK145083.1 | - | 10 - 11 | 1302 | 68 | |
BC028341.1 | - | 10 - 11 | 1309 | 68 |