Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tyk2
Entrez Gene ID
54721
Gene Name
tyrosine kinase 2
Gene Aliases
JTK1
Chromosome Location
Chr.9: 21104068 - 21131275 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205312.1 NP_001192241.1 22 - 23 3530 85
NM_018793.2 NP_061263.2 22 - 23 3461 85
GenBank mRNA AF173032.1 - 22 - 23 3396 85
AK154438.1 - 22 - 23 3488 85
AK155923.1 - 22 - 23 3373 85
AK159303.1 - 22 - 23 3430 85
AK167726.1 - 22 - 23 3440 85
AK170107.1 - 22 - 23 3400 85
AK170940.1 - 22 - 23 3399 85
AK171609.1 - 22 - 23 3373 85
BC019789.1 - 6 - 7 877 85
BC053021.1 - 16 - 17 3101 85
BC094240.1 - 21 - 22 3357 85

View Full Product Details