Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt7
Entrez Gene ID
54525
Gene Name
synaptotagmin VII
Gene Aliases
AI851541, B230112P13Rik
Chromosome Location
Chr.19: 10388902 - 10453181 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018801.3 NP_061271.1 8 - 9 1500 59
NM_173067.3 NP_775090.1 9 - 10 1632 59
NM_173068.2 NP_775091.2 10 - 11 1992 59
GenBank mRNA AB026804.1 - 8 - 9 1107 59
AB075900.1 - 9 - 10 1239 59
AB075901.1 - 11 - 12 1452 59
AK075696.1 - 7 - 8 1300 59
BC105660.1 - 8 - 9 1106 59
BC139806.1 - 8 - 9 1107 59

View Full Product Details