The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019505.3 | NP_062378.1 | 1 - 2 | 820 | 63 |
GenBank mRNA | AF136744.1 | - | 1 - 2 | 457 | 63 |
AK044596.1 | - | - | 1556 | 63 | |
AK049335.1 | - | 1 - 2 | 620 | 63 | |
AK147841.1 | - | 1 - 2 | 821 | 63 | |
BB635763.1 | - | 1 - 2 | 224 | 63 | |
BC055867.1 | - | 1 - 2 | 753 | 63 |