Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il17rb
Entrez Gene ID
50905
Gene Name
interleukin 17 receptor B
Gene Aliases
Evi27, IL-17ER, IL-17Rh1, IL17RH1, Il17br
Chromosome Location
Chr.14: 29995506 - 30008896 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019583.3 NP_062529.2 6 - 7 637 108
GenBank mRNA AF208108.1 - 6 - 7 550 108
AF208109.1 - 6 - 7 1176 108
AK045627.1 - 6 - 7 638 108
AK050139.1 - 6 - 7 558 108
BC026546.1 - 6 - 7 537 108

View Full Product Details