The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019583.3 | NP_062529.2 | 9 - 10 | 942 | 90 |
GenBank mRNA | AF208108.1 | - | 9 - 10 | 855 | 90 |
AF208109.1 | - | 9 - 10 | 1481 | 90 | |
AK045627.1 | - | 9 - 10 | 943 | 90 | |
AK050139.1 | - | 9 - 10 | 863 | 90 | |
BC026546.1 | - | 9 - 10 | 842 | 90 |