Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pfkp
Entrez Gene ID
56421
Gene Name
phosphofructokinase, platelet
Gene Aliases
1200015H23Rik, 9330125N24Rik, ATP-PFK, PFK-C, PFK-P
Chromosome Location
Chr.13: 6579874 - 6648789 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291071.1 NP_001278000.1 21 - 22 2311 63
NM_019703.4 NP_062677.1 21 - 22 2311 63
GenBank mRNA AF123533.1 - 21 - 22 2276 63
AK004790.1 - 21 - 22 2265 63
AK047303.1 - 3 - 4 310 63
AK049841.1 - 21 - 22 2265 63
AK076803.1 - 21 - 22 2266 63
AK078254.1 - 21 - 22 2266 63
AK098162.1 - 21 - 22 2265 63
AK152670.1 - 21 - 22 2266 63
AK154125.1 - 21 - 22 2266 63
AK170624.1 - 21 - 22 2266 63
AK171062.1 - 21 - 22 2266 63
AY779275.1 - 21 - 22 2228 63
AY779276.1 - 21 - 22 2228 63
BC006926.1 - 21 - 22 2250 63
CT010268.1 - 21 - 22 2219 63
Y19008.1 - 21 - 22 2268 63

View Full Product Details