Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gabrq
Entrez Gene ID
57249
Gene Name
gamma-aminobutyric acid (GABA) A receptor, subunit theta
Gene Aliases
-
Chromosome Location
Chr.X: 72825163 - 72842607 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290435.1 NP_001277364.1 5 - 6 892 108
NM_020488.2 NP_065234.1 5 - 6 892 108
GenBank mRNA AF189260.1 - 5 - 6 634 108
AK038859.1 - 5 - 6 803 108
AK156674.1 - 5 - 6 893 108
BC119074.1 - 5 - 6 632 108
BC119076.1 - 5 - 6 632 108

View Full Product Details