The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163753.1 | NP_001157225.1 | 5 - 6 | 774 | 65 |
NM_021411.4 | NP_067386.3 | 5 - 6 | 386 | 65 | |
GenBank mRNA | AB232633.1 | - | 5 - 6 | 365 | 65 |
AF233582.1 | - | 5 - 6 | 365 | 65 | |
AK039716.1 | - | 5 - 6 | 380 | 65 | |
AK040216.1 | - | 5 - 6 | 386 | 65 | |
AK045995.1 | - | 5 - 6 | 377 | 65 | |
AK046260.1 | - | 5 - 6 | 775 | 65 | |
AK050483.1 | - | 5 - 6 | 380 | 65 | |
BC050757.1 | - | 5 - 6 | 386 | 65 | |
BC128020.1 | - | 5 - 6 | 384 | 65 |