The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021537.3 | NP_067512.3 | 3 - 4 | 501 | 76 |
GenBank mRNA | AF004934.1 | - | 2 - 3 | 339 | 76 |
AK148041.1 | - | 3 - 4 | 475 | 76 | |
AK152917.1 | - | 3 - 4 | 444 | 76 | |
BC029875.1 | - | 3 - 4 | 403 | 76 | |
BC052913.1 | - | 3 - 4 | 436 | 76 | |
BC071218.1 | - | 3 - 4 | 496 | 76 |