The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022331.1 | NP_071726.1 | 7 - 8 | 1099 | 69 |
GenBank mRNA | AB034991.1 | - | 7 - 8 | 1099 | 69 |
AJ320519.1 | - | 1 - 2 | 106 | 69 | |
AK081688.1 | - | 7 - 8 | 1103 | 69 | |
AK159500.1 | - | 7 - 8 | 1105 | 69 | |
AK165832.1 | - | 7 - 8 | 1105 | 69 | |
AK172874.1 | - | 7 - 8 | 1092 | 69 | |
BC013523.1 | - | 7 - 8 | 1091 | 69 |