The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024226.4 | NP_077188.1 | 2 - 3 | 528 | 90 |
NM_194051.3 | NP_918940.1 | 4 - 5 | 2815 | 90 | |
NM_194052.3 | NP_918941.1 | 2 - 3 | 1004 | 90 | |
NM_194053.3 | NP_918942.1 | 3 - 4 | 1061 | 90 | |
NM_194054.3 | NP_918943.1 | 4 - 5 | 3422 | 90 | |
GenBank mRNA | AB073672.1 | - | 2 - 3 | 1380 | 90 |
AF326337.1 | - | 2 - 3 | 308 | 90 | |
AK034902.1 | - | 2 - 3 | 948 | 90 | |
AK212390.1 | - | 2 - 3 | 231 | 90 | |
AK220511.1 | - | 4 - 5 | 3382 | 90 | |
AY102280.1 | - | 4 - 5 | 2815 | 90 | |
AY102281.1 | - | 2 - 3 | 961 | 90 | |
AY102282.1 | - | 3 - 4 | 1018 | 90 | |
AY102283.1 | - | 2 - 3 | 521 | 90 | |
AY102284.1 | - | 4 - 5 | 3379 | 90 | |
AY114152.1 | - | 4 - 5 | 3380 | 90 | |
AY114153.1 | - | 3 - 4 | 835 | 90 | |
BC032192.1 | - | 2 - 3 | 1674 | 90 | |
BC032272.1 | - | 2 - 3 | 2528 | 90 | |
BC056373.1 | - | 4 - 5 | 3250 | 90 | |
BU848611.1 | - | 2 - 3 | 367 | 90 |