The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172481.1 | NP_001165952.1 | 7 - 8 | 1205 | 83 |
NM_025711.3 | NP_079987.2 | 7 - 8 | 1222 | 83 | |
GenBank mRNA | AF316825.1 | - | 6 - 7 | 942 | 83 |
AK014504.1 | - | 7 - 8 | 1178 | 83 | |
AK085920.1 | - | 7 - 8 | 1178 | 83 | |
BC034888.1 | - | 7 - 8 | 1124 | 83 | |
BC145904.1 | - | 7 - 8 | 1120 | 83 | |
BC145930.1 | - | 7 - 8 | 1103 | 83 |