The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030724.3 | NP_109649.1 | 1 - 2 | 349 | 75 |
GenBank mRNA | AF236636.1 | - | 1 - 2 | 354 | 75 |
AK051894.1 | - | 1 - 2 | 119 | 75 | |
AK053745.1 | - | 1 - 2 | 309 | 75 | |
AK077517.1 | - | 1 - 2 | 284 | 75 | |
AK082837.1 | - | 1 - 2 | 312 | 75 | |
AK155939.1 | - | 1 - 2 | 313 | 75 | |
AK168318.1 | - | 1 - 2 | 327 | 75 | |
BC023789.1 | - | 1 - 2 | 313 | 75 |