The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031884.1 | NP_114090.1 | 10 - 11 | 1516 | 65 |
GenBank mRNA | AF312713.2 | - | 10 - 11 | 1604 | 65 |
AK149569.1 | - | 10 - 11 | 1487 | 65 | |
AY195873.1 | - | 10 - 11 | 1604 | 65 | |
BC106766.2 | - | 10 - 11 | 1493 | 65 |