Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Copb1
Entrez Gene ID
70349
Gene Name
coatomer protein complex, subunit beta 1
Gene Aliases
2610019B04Rik
Chromosome Location
Chr.7: 114215559 - 114254680 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033370.3 NP_203534.1 13 - 14 1752 80
GenBank mRNA AF231925.1 - 12 - 13 1691 80
AK135168.1 - 13 - 14 1784 80
AK149828.1 - 13 - 14 1770 80
AK161544.1 - 13 - 14 1779 80
AK172214.1 - 13 - 14 1754 80
BC030837.1 - 13 - 14 1752 80

View Full Product Details