The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033444.2 | NP_254279.1 | 1 - 2 | 245 | 61 |
GenBank mRNA | AK088809.1 | - | 1 - 2 | 56 | 61 |
AK150382.1 | - | 1 - 2 | 523 | 61 | |
AK150611.1 | - | 1 - 2 | 234 | 61 | |
AK153509.1 | - | 1 - 2 | 523 | 61 | |
AK167765.1 | - | 1 - 2 | 251 | 61 | |
AK168262.1 | - | 1 - 2 | 65 | 61 | |
BC004658.1 | - | 1 - 2 | 245 | 61 |