The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164311.1 | NP_001157783.1 | 2 - 3 | 384 | 79 |
NM_053083.3 | NP_444313.3 | 2 - 3 | 384 | 79 | |
GenBank mRNA | AF338440.1 | - | 1 - 2 | 279 | 79 |
AK014331.1 | - | 2 - 3 | 384 | 79 | |
BC068307.1 | - | 2 - 3 | 326 | 79 |