Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sfxn3
Entrez Gene ID
94280
Gene Name
sideroflexin 3
Gene Aliases
-
Chromosome Location
Chr.19: 45047508 - 45056383 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178012.1 NP_001171483.1 8 - 9 1037 141
NM_053197.4 NP_444427.1 9 - 10 1136 141
GenBank mRNA AF325262.1 - 9 - 10 900 141
AK034514.1 - 9 - 10 853 141
AK089985.1 - 8 - 9 798 141
BC012208.1 - 9 - 10 1038 141
BC013146.1 - 3 - 4 127 141

View Full Product Details