Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rasa2
Entrez Gene ID
114713
Gene Name
RAS p21 protein activator 2
Gene Aliases
5430433H21Rik, AA517451, AU023900, GAP1m
Chromosome Location
Chr.9: 96539300 - 96631615 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053268.2 NP_444498.2 15 - 16 1593 104
GenBank mRNA AB056433.1 - 15 - 16 1586 104
AK034434.1 - 15 - 16 1593 104
AK035664.1 - 6 - 7 616 104
AK082530.1 - 5 - 6 2987 104
AK154499.1 - 15 - 16 1610 104
AK166654.1 - 15 - 16 1593 104

View Full Product Details