Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scgb3a1
Entrez Gene ID
68662
Gene Name
secretoglobin, family 3A, member 1
Gene Aliases
HIN-1, LuLeu2, Pnsp2, UGRP2, Utgrp2
Chromosome Location
Chr.11: 49663595 - 49665118 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054037.2 NP_473378.1 1 - 2 124 59
GenBank mRNA AF313456.1 - 1 - 2 109 59
AF436840.1 - 1 - 2 93 59
AV578532.1 - 1 - 2 124 59
BC086910.1 - 1 - 2 106 59

View Full Product Details