The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080637.3 | NP_542368.2 | 5 - 6 | 669 | 118 |
GenBank mRNA | AF343565.1 | - | 4 - 5 | 558 | 118 |
AK006111.1 | - | 3 - 4 | 552 | 118 | |
AK131722.1 | - | 5 - 6 | 670 | 118 | |
AK138562.1 | - | - | 344 | 118 | |
BC049625.1 | - | 4 - 5 | 562 | 118 |