Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vps26a
Entrez Gene ID
30930
Gene Name
vacuolar protein sorting 26 homolog A (yeast)
Gene Aliases
AA407240, HB58, Vps26
Chromosome Location
Chr.10: 62454843 - 62486805 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113355.1 NP_001106826.1 4 - 5 607 115
NM_133672.3 NP_598433.1 4 - 5 534 115
GenBank mRNA AK028096.1 - 4 - 5 608 115
AK086851.1 - - 430 115
AK147989.1 - 4 - 5 475 115
AK149859.1 - 4 - 5 459 115
AK150244.1 - 4 - 5 432 115
AK151360.1 - 4 - 5 444 115
AK152424.1 - 4 - 5 478 115
AK159783.1 - 4 - 5 484 115
AK165853.1 - 4 - 5 478 115
AK167592.1 - 4 - 5 489 115
AK168213.1 - 4 - 5 534 115
AK168537.1 - 4 - 5 489 115
BC007148.1 - 4 - 5 455 115
S41204.1 - 4 - 5 481 115

View Full Product Details