Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dgkg
Entrez Gene ID
110197
Gene Name
diacylglycerol kinase, gamma
Gene Aliases
2900055E17Rik, 90kDa, AI854428, Dagk3, E430001K23Rik, mKIAA4131
Chromosome Location
Chr.16: 22466568 - 22657299 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138650.2 NP_619591.1 18 - 19 2047 75
GenBank mRNA AK083258.1 - 17 - 18 1923 75
AK087972.1 - 18 - 19 2258 75
AK220294.1 - 18 - 19 1989 75
BC015278.1 - 17 - 18 1620 75

View Full Product Details