Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc6a13
Entrez Gene ID
14412
Gene Name
solute carrier family 6 (neurotransmitter transporter, GABA), member 13
Gene Aliases
GAT3, Gabt3, Gat2
Chromosome Location
Chr.6: 121300296 - 121337724 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144512.2 NP_653095.1 14 - 15 1726 84
GenBank mRNA AK149557.1 - 14 - 15 1734 84
BC021606.1 - 5 - 6 584 84
BC023117.1 - 14 - 15 1714 84
BC029637.1 - 14 - 15 1726 84

View Full Product Details