The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110145.1 | NP_001103615.1 | 7 - 8 | 803 | 71 |
NM_009278.4 | NP_033304.1 | 7 - 8 | 830 | 71 | |
GenBank mRNA | AK032940.1 | - | 7 - 8 | 747 | 71 |
AK088650.1 | - | 7 - 8 | 786 | 71 | |
AK088677.1 | - | 7 - 8 | 737 | 71 | |
AK134864.1 | - | 7 - 8 | 768 | 71 | |
AK141487.1 | - | 7 - 8 | 725 | 71 | |
AK168828.1 | - | 7 - 8 | 753 | 71 | |
BC003820.1 | - | 7 - 8 | 690 | 71 | |
BC106150.1 | - | 7 - 8 | 683 | 71 | |
L00993.1 | - | 7 - 8 | 766 | 71 |