The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166480.1 | NP_001159952.1 | 2 - 3 | 395 | 65 |
NM_009279.3 | NP_033305.1 | 2 - 3 | 277 | 65 | |
GenBank mRNA | AK003044.1 | - | 2 - 3 | 257 | 65 |
AK007921.1 | - | 2 - 3 | 233 | 65 | |
AK013723.1 | - | 2 - 3 | 239 | 65 | |
AK160089.1 | - | 2 - 3 | 243 | 65 | |
BC008172.1 | - | 2 - 3 | 198 | 65 | |
BC012971.1 | - | 1 - 2 | 1778 | 65 | |
BC019419.1 | - | 1 - 2 | 1728 | 65 | |
BI989805.1 | - | 2 - 3 | 395 | 65 | |
BQ747160.1 | - | 2 - 3 | 180 | 65 | |
CJ144362.1 | - | 2 - 3 | 282 | 65 | |
X90582.1 | - | 2 - 3 | 358 | 65 |