Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tkt
Entrez Gene ID
21881
Gene Name
transketolase
Gene Aliases
p68
Chromosome Location
Chr.14: 30549131 - 30574724 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009388.6 NP_033414.1 4 - 5 581 66
GenBank mRNA AF195533.1 - 3 - 4 247 66
AK002627.1 - 4 - 5 534 66
AK012794.1 - 4 - 5 536 66
AK030446.1 - 4 - 5 564 66
AK140965.1 - 4 - 5 565 66
AK144146.1 - 4 - 5 536 66
AK145677.1 - 4 - 5 534 66
AK146157.1 - 4 - 5 534 66
AK150139.1 - 4 - 5 534 66
AK150769.1 - 4 - 5 530 66
AK150844.1 - 4 - 5 530 66
AK150856.1 - 4 - 5 530 66
AK150901.1 - 4 - 5 534 66
AK151916.1 - 4 - 5 530 66
AK151969.1 - 4 - 5 530 66
AK152245.1 - 4 - 5 530 66
AK152460.1 - 4 - 5 550 66
AK152886.1 - 4 - 5 530 66
AK159922.1 - 4 - 5 534 66
AK166763.1 - 4 - 5 568 66
AK167084.1 - 4 - 5 568 66
AK177246.1 - 3 - 4 243 66
BC055336.1 - 4 - 5 508 66
U05809.1 - 4 - 5 529 66

View Full Product Details