The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009410.2 | NP_033436.1 | 18 - 19 | 3099 | 62 |
GenBank mRNA | AB006074.1 | - | 18 - 19 | 3100 | 62 |
BC099689.1 | - | 18 - 19 | 2914 | 62 |