The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009476.2 | NP_033502.2 | 1 - 2 | 140 | 81 |
GenBank mRNA | BC104260.1 | - | 1 - 2 | 140 | 81 |
BC104261.1 | - | 1 - 2 | 140 | 81 | |
CX565093.1 | - | 1 - 2 | 350 | 81 | |
DQ358978.1 | - | 1 - 2 | 79 | 81 | |
U08030.1 | - | 1 - 2 | 138 | 81 |