The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010865.3 | NP_034995.3 | 1 - 2 | 682 | 68 |
GenBank mRNA | AB013592.1 | - | 1 - 2 | 666 | 68 |
AF039869.1 | - | 1 - 2 | 663 | 68 | |
AK144395.1 | - | 1 - 2 | 678 | 68 | |
BC125329.1 | - | 1 - 2 | 627 | 68 | |
BC137606.1 | - | 1 - 2 | 627 | 68 |