The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159717.1 | NP_001153189.1 | 4 - 5 | 313 | 78 |
NM_001159718.1 | NP_001153190.1 | 3 - 4 | 286 | 78 | |
NM_001159719.1 | NP_001153191.1 | 4 - 5 | 361 | 78 | |
NM_010891.2 | NP_035021.1 | 4 - 5 | 317 | 78 | |
GenBank mRNA | AK028072.1 | - | 4 - 5 | 299 | 78 |
AK146616.1 | - | 4 - 5 | 314 | 78 | |
AK151591.1 | - | 4 - 5 | 301 | 78 | |
AK169852.1 | - | 3 - 4 | 336 | 78 | |
AK171331.1 | - | 4 - 5 | 296 | 78 | |
BB849650.1 | - | 4 - 5 | 287 | 78 | |
BC138636.1 | - | 3 - 4 | 254 | 78 | |
BC138637.1 | - | 3 - 4 | 254 | 78 | |
BY330456.1 | - | 3 - 4 | 262 | 78 | |
D49382.2 | - | 4 - 5 | 272 | 78 |