Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Musk
Entrez Gene ID
18198
Gene Name
muscle, skeletal, receptor tyrosine kinase
Gene Aliases
Mdk4, Mlk, Nsk1, Nsk2, Nsk3
Chromosome Location
Chr.4: 58285640 - 58377848 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037127.2 NP_001032204.2 13 - 14 1802 77
NM_001037128.1 NP_001032205.1 13 - 14 1757 77
NM_001037129.1 NP_001032206.1 12 - 13 1733 77
NM_001037130.1 NP_001032207.1 11 - 12 1703 77
NM_010944.2 NP_035074.2 12 - 13 1727 77
GenBank mRNA AB691772.1 - 11 - 12 1553 77
AY955296.1 - 13 - 14 1652 77
AY955297.1 - 13 - 14 1607 77
AY955298.1 - 12 - 13 1583 77
AY956403.1 - 12 - 13 1577 77
AY956404.1 - 11 - 12 1553 77
BC100342.1 - 12 - 13 1657 77
U37708.1 - 11 - 12 1701 77
U37709.1 - 12 - 13 1725 77
X86444.1 - 11 - 12 1631 77

View Full Product Details