Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phex
Entrez Gene ID
18675
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
Gy, HPDR, HPDR1, Hyp, PEX
Chromosome Location
Chr.X: 157162075 - 157416551 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011077.2 NP_035207.1 6 - 7 1263 88
GenBank mRNA AK158481.1 - 6 - 7 1290 88
BC137925.1 - 6 - 7 1010 88
BC137926.1 - 6 - 7 1010 88
EF194891.1 - 6 - 7 797 88
U49908.1 - 6 - 7 1255 88
U73910.1 - 6 - 7 1263 88
U75646.1 - 6 - 7 844 88

View Full Product Details