Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pou2af1
Entrez Gene ID
18985
Gene Name
POU domain, class 2, associating factor 1
Gene Aliases
BOB.1, Bob-1, Bob1, OBF-1, OBF.1, OCA-B, OCAB
Chromosome Location
Chr.9: 51213690 - 51240079 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011136.2 NP_035266.1 2 - 3 249 99
GenBank mRNA AK088587.1 - 2 - 3 249 99
AK135080.1 - 2 - 3 250 99
BC058611.1 - 2 - 3 230 99
U43788.1 - 2 - 3 249 99

View Full Product Details