The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011203.2 | NP_035333.2 | 1 - 2 | 145 | 106 |
GenBank mRNA | AK154107.1 | - | 1 - 2 | 237 | 106 |
BC051980.1 | - | 1 - 2 | 145 | 106 | |
X63440.1 | - | 1 - 2 | 146 | 106 |