The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159385.1 | NP_001152857.1 | 2 - 3 | 110 | 76 |
NM_011550.3 | NP_035680.3 | 2 - 3 | 272 | 76 | |
GenBank mRNA | AF213671.1 | - | 2 - 3 | 74 | 76 |
AF213672.1 | - | 2 - 3 | 236 | 76 | |
AK177451.1 | - | 1 - 2 | 64 | 76 | |
BC031380.1 | - | 2 - 3 | 252 | 76 | |
BC038004.1 | - | 2 - 3 | 252 | 76 | |
BC080768.1 | - | 1 - 2 | 1076 | 77 | |
CK620266.1 | - | 2 - 3 | 114 | 76 | |
U43548.1 | - | 2 - 3 | 232 | 76 | |
U43607.1 | - | 2 - 3 | 243 | 76 |