The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011563.5 | NP_035693.3 | 1 - 2 | 118 | 64 |
GenBank mRNA | AK008433.1 | - | 1 - 2 | 123 | 64 |
AK010653.1 | - | 1 - 2 | 93 | 64 | |
AK011963.1 | - | - | 66 | 64 | |
AK088280.1 | - | 1 - 2 | 137 | 64 | |
BC002034.1 | - | 1 - 2 | 126 | 64 | |
BC081454.1 | - | 1 - 2 | 37 | 64 | |
U20611.1 | - | 1 - 2 | 80 | 64 | |
U51679.1 | - | 1 - 2 | 55 | 64 |