Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thbs2
Entrez Gene ID
21826
Gene Name
thrombospondin 2
Gene Aliases
TSP2, Thbs-2
Chromosome Location
Chr.17: 14665500 - 14694262 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011581.3 NP_035711.2 11 - 12 2172 100
GenBank mRNA AK132508.1 - 11 - 12 2003 100
AK142552.1 - 11 - 12 2146 100
BC053702.1 - 11 - 12 1974 100
L07803.1 - 11 - 12 2000 100
M64866.1 - 11 - 12 2000 100

View Full Product Details