Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnnt3
Entrez Gene ID
21957
Gene Name
troponin T3, skeletal, fast
Gene Aliases
fTnT
Chromosome Location
Chr.7: 142498777 - 142516213 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163664.1 NP_001157136.1 14 - 15 603 61
NM_001163665.1 NP_001157137.1 14 - 15 603 61
NM_001163666.1 NP_001157138.1 12 - 13 576 61
NM_001163667.1 NP_001157139.1 13 - 14 591 61
NM_001163668.1 NP_001157140.1 12 - 13 552 61
NM_001163669.1 NP_001157141.1 13 - 14 564 61
NM_001163670.1 NP_001157142.1 10 - 11 519 61
NM_011620.3 NP_035750.2 11 - 12 531 61
GenBank mRNA AI746973.1 - 12 - 13 571 61
AK003147.1 - 10 - 11 486 61
BC003747.1 - 14 - 15 599 61
CB196352.1 - 4 - 5 355 61
CK791812.1 - 3 - 4 241 61
DV049834.1 - 11 - 12 578 61
L48988.1 - 8 - 9 422 61
L48989.1 - 8 - 9 422 61
L48990.1 - 11 - 12 467 61
L48991.1 - 8 - 9 449 61
L48992.1 - 10 - 11 455 61
L48993.1 - 10 - 11 455 61
L49018.1 - 8 - 9 449 61
L49466.1 - 13 - 14 591 61
L49467.1 - 14 - 15 603 61
L49468.1 - 12 - 13 576 61
L49469.1 - 12 - 13 576 61
L49470.1 - 13 - 14 564 61
L49471.1 - 12 - 13 573 61
L49472.1 - 12 - 13 552 61
U77779.1 - 11 - 12 524 61

View Full Product Details